missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrosodiphenylamine 97.0+%, TCI America™

Catalog No. n026325g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N026325G Supplier TCI America Supplier No. N026325G
Only null left

Chemical Identifiers

CAS 156-10-5
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002062
InChI Key OIJHFHYPXWSVPF-UHFFFAOYSA-N
Synonym 4-nitrosodiphenylamine, p-nitrosodiphenylamine, 4-nitroso-n-phenylbenzenamine, p-nitroso-n-phenylaniline, diphenylamine, 4-nitroso, naugard tkb, benzenamine, 4-nitroso-n-phenyl, p-nitrosodifenylamin, n-phenyl-p-nitrosoaniline, p-phenylaminonitrosobenzene
PubChem CID 9074
ChEBI CHEBI:82509
IUPAC Name 4-nitroso-N-phenylaniline
SMILES O=NC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 198.23
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrosodiphenylamine
EINECSNumber (3)-0463
RTECSNumber JK0175000
TSCA Yes