CAS RN 156-10-5
CAS RN 156-10-5
IUPAC Name:
4-nitroso-N-phenylaniline
Synonyms:
4-nitroso-N-phenylbenzenamine
Molecular Weight (g/mol):
198.23
Molecular Formula:
C12H10N2O
InChi Key:
OIJHFHYPXWSVPF-UHFFFAOYSA-N
SMILES:
O=NC1=CC=C(NC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Nitrosodiphenylamine 97.0+%, TCI America™
CAS: 156-10-5 Molecular Formula: C12H10N2O Molecular Weight (g/mol): 198.23 MDL Number: MFCD00002062 InChI Key: OIJHFHYPXWSVPF-UHFFFAOYSA-N Synonym: 4-nitrosodiphenylamine,p-nitrosodiphenylamine,4-nitroso-n-phenylbenzenamine,p-nitroso-n-phenylaniline,diphenylamine, 4-nitroso,naugard tkb,benzenamine, 4-nitroso-n-phenyl,p-nitrosodifenylamin,n-phenyl-p-nitrosoaniline,p-phenylaminonitrosobenzene PubChem CID: 9074 ChEBI: CHEBI:82509 IUPAC Name: 4-nitroso-N-phenylaniline SMILES: O=NC1=CC=C(NC2=CC=CC=C2)C=C1