missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrosalicylic Acid 98.0+%, TCI America™
$270.29 - $1,081.16
Chemical Identifiers
| CAS | 619-19-2 |
|---|---|
| Molecular Formula | C7H5NO5 |
| Molecular Weight (g/mol) | 183.12 |
| MDL Number | MFCD00071505 |
| InChI Key | UKWUOTZGXIZAJC-UHFFFAOYSA-N |
| Synonym | 4-nitrosalicylic acid, 2-hydroxy-4-nitrobenzoic acid, benzoic acid, 2-hydroxy-4-nitro, 4-nitro-2-hydroxybenzoic acid, p-nitrosalicylic acid, salicylic acid, 4-nitro, 4-nitro-salicylsaure, 4-nitro-salicylsaure german, 2-hydroxy-4-nitro-benzoic acid, p-nitrosalicylsaure |
| PubChem CID | 69266 |
| IUPAC Name | 2-hydroxy-4-nitrobenzoic acid |
| SMILES | OC(=O)C1=CC=C(C=C1O)[N+]([O-])=O |
Chemical Identifiers
| 619-19-2 | |
| 183.12 | |
| UKWUOTZGXIZAJC-UHFFFAOYSA-N | |
| 69266 | |
| OC(=O)C1=CC=C(C=C1O)[N+]([O-])=O |
| C7H5NO5 | |
| MFCD00071505 | |
| 4-nitrosalicylic acid, 2-hydroxy-4-nitrobenzoic acid, benzoic acid, 2-hydroxy-4-nitro, 4-nitro-2-hydroxybenzoic acid, p-nitrosalicylic acid, salicylic acid, 4-nitro, 4-nitro-salicylsaure, 4-nitro-salicylsaure german, 2-hydroxy-4-nitro-benzoic acid, p-nitrosalicylsaure | |
| 2-hydroxy-4-nitrobenzoic acid |
Specifications
| 619-19-2 | |
| C7H5NO5 | |
| 5 g | |
| UKWUOTZGXIZAJC-UHFFFAOYSA-N | |
| 2-hydroxy-4-nitrobenzoic acid | |
| 69266 | |
| ≥98.0% (T) | |
| 4-Nitrosalicylic Acid |
| 234°C | |
| MFCD00071505 | |
| 4-nitrosalicylic acid, 2-hydroxy-4-nitrobenzoic acid, benzoic acid, 2-hydroxy-4-nitro, 4-nitro-2-hydroxybenzoic acid, p-nitrosalicylic acid, salicylic acid, 4-nitro, 4-nitro-salicylsaure, 4-nitro-salicylsaure german, 2-hydroxy-4-nitro-benzoic acid, p-nitrosalicylsaure | |
| OC(=O)C1=CC=C(C=C1O)[N+]([O-])=O | |
| 183.12 | |
| 183.12 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VO5300100
TSCA : Yes