missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyrrole-2-carboxylic Acid Hydrate 99.0+%, TCI America™

Catalog No. N05091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05091G 1 g
N05095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05091G Supplier TCI America Supplier No. N05091G

Chemical Identifiers

CAS 5930-93-8
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD00059928
InChI Key SNWGRRCTCGASCN-UHFFFAOYSA-N
PubChem CID 7016560
SMILES C1=C(NC=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point 217°C
Quantity 1 g
Formula Weight 156.10
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitropyrrole-2-carboxylic Acid Hydrate