missing translation for 'onlineSavingsMsg'
Learn More

4-Nitropyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133882
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0589100MG 100 mg
N05891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0589100MG Supplier TCI America Supplier No. N0589100MG

Chemical Identifiers

CAS 13509-19-8
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.108
MDL Number MFCD00191562
InChI Key USTVSOYPMQZLSA-UHFFFAOYSA-N
Synonym 4-Nitropicolinic Acid
PubChem CID 2762825
SMILES C1=CN=C(C=C1[N+](=O)[O-])C(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 168.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitropyridine-2-carboxylic Acid