accessibility menu, dialog, popup

UserName

NNOOOHO

CAS RN 13509-19-8

IUPAC Name: 4-nitropyridine-2-carboxylic acid
Synonyms: 4-nitropyridine-2-carboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: USTVSOYPMQZLSA-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results