missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic Hydrazide 98.0+%, TCI America™

Catalog No. p-7133979
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07995G 5 g
N079925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07995G Supplier TCI America Supplier No. N07995G
Only null left

Chemical Identifiers

CAS 3682-19-7
Molecular Formula C8H5N3O4
Molecular Weight (g/mol) 207.15
MDL Number MFCD00006891
InChI Key XQFIOLKLHNOFCA-UHFFFAOYSA-N
Synonym 4-Nitrophthalhydrazide, 2,3-Dihydro-6-nitro-1,4-phthalazinedione
PubChem CID 3543259
IUPAC Name 6-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione
SMILES [O-][N+](=O)C1=CC=C2C(=O)NNC(=O)C2=C1

Specifications

Melting Point 300°C
Color Yellow
Quantity 5 g
Formula Weight 207.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalic Hydrazide
RTECSNumber TH8895000
TSCA No