accessibility menu, dialog, popup

UserName

ONOONHNHO

CAS RN 3682-19-7

IUPAC Name: 6-nitro-1,2,3,4-tetrahydrophthalazine-1,4-dione
Synonyms: 6-nitro-2,3-dihydrophthalazine-1,4-dione
Molecular Weight (g/mol): 207.15
Molecular Formula: C8H5N3O4
InChi Key: XQFIOLKLHNOFCA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C(=O)NNC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results