missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic acid, 97%

Catalog No. AAA1036818
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1036818 50 g
AAA1036806 5 g
AAA1036814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1036818 Supplier Thermo Scientific Chemicals Supplier No. A1036818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 610-27-5 | C8H5NO6 | 211.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 610-27-5
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007252
InChI Key SLBQXWXKPNIVSQ-UHFFFAOYSA-N
Synonym 4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8
PubChem CID 69121
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 160°C to 165°C
Odor Odorless
Quantity 50 g
Beilstein 1882262
Formula Weight 211.13
Percent Purity 97%
Chemical Name or Material 4-Nitrophthalic acid
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-215-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only