missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenylsulfur Pentafluoride 96.0+%, TCI America™
$46.60 - $96.94
Chemical Identifiers
| CAS | 2613-27-6 |
|---|---|
| Molecular Formula | C6H4F5NO2S |
| Molecular Weight (g/mol) | 249.155 |
| MDL Number | MFCD00221619 |
| InChI Key | AGNCKMHGYZKMLN-UHFFFAOYSA-N |
| Synonym | 4-nitrophenylsulfur pentafluoride, 4-nitrophenylsulphur pentafluoride, 4-pentafluorosulfur-nitrobenzene, 1-nitro-4-pentafluorosulfanyl benzene, 1-nitro-4-pentafluoro-??-sulfanyl benzene, acmc-209gok, pentafluoro-4-nitrophenyl, 4-pentafluorothio nitrobenzene, 4-nitrophenylsulfurpentafluoride, 4-nitrophenylpentafluorosulfur vi |
| PubChem CID | 2775740 |
| IUPAC Name | pentafluoro-(4-nitrophenyl)-$l^{6}-sulfane |
| SMILES | C1=CC(=CC=C1[N+](=O)[O-])S(F)(F)(F)(F)F |
Chemical Identifiers
| 2613-27-6 | |
| 249.155 | |
| AGNCKMHGYZKMLN-UHFFFAOYSA-N | |
| 2775740 | |
| C1=CC(=CC=C1[N+](=O)[O-])S(F)(F)(F)(F)F |
| C6H4F5NO2S | |
| MFCD00221619 | |
| 4-nitrophenylsulfur pentafluoride, 4-nitrophenylsulphur pentafluoride, 4-pentafluorosulfur-nitrobenzene, 1-nitro-4-pentafluorosulfanyl benzene, 1-nitro-4-pentafluoro-??-sulfanyl benzene, acmc-209gok, pentafluoro-4-nitrophenyl, 4-pentafluorothio nitrobenzene, 4-nitrophenylsulfurpentafluoride, 4-nitrophenylpentafluorosulfur vi | |
| pentafluoro-(4-nitrophenyl)-$l^{6}-sulfane |
Specifications
| 2613-27-6 | |
| Yellow | |
| C6H4F5NO2S | |
| 1 g | |
| 4-nitrophenylsulfur pentafluoride, 4-nitrophenylsulphur pentafluoride, 4-pentafluorosulfur-nitrobenzene, 1-nitro-4-pentafluorosulfanyl benzene, 1-nitro-4-pentafluoro-??-sulfanyl benzene, acmc-209gok, pentafluoro-4-nitrophenyl, 4-pentafluorothio nitrobenzene, 4-nitrophenylsulfurpentafluoride, 4-nitrophenylpentafluorosulfur vi | |
| C1=CC(=CC=C1[N+](=O)[O-])S(F)(F)(F)(F)F | |
| 249.155 | |
| 249.16 | |
| Crystalline Powder |
| 39°C | |
| 77°C | |
| MFCD00221619 | |
| 2811 | |
| AGNCKMHGYZKMLN-UHFFFAOYSA-N | |
| pentafluoro-(4-nitrophenyl)-$l^{6}-sulfane | |
| 2775740 | |
| ≥96.0% (GC) | |
| 4-Nitrophenylsulfur Pentafluoride |
Safety and Handling
TSCA : No