missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7133693
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02335G 5 g
N023325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02335G Supplier TCI America Supplier No. N02335G

Chemical Identifiers

CAS 636-99-7
Molecular Formula C6H8ClN3O2
Molecular Weight (g/mol) 189.599
MDL Number MFCD00040592
InChI Key ZWWXDCOPVYATOQ-UHFFFAOYSA-N
Synonym 4-nitrophenylhydrazine hydrochloride, 4-nitrophenyl hydrazine hydrochloride, 4-nitrophenyhydrazine hydrochloride, 4-nitrophenylhydrazinehydrochloride, 4-nitrophenyl hydrazine, hcl, p-nitrophenylhydrazine hcl, nitrophenylhydrazine hydrochloride, hydrazine, 4-nitrophenyl-, monohydrochloride, p-nitrophenylhydrazine hydrochloride
PubChem CID 5463401
IUPAC Name (4-nitrophenyl)hydrazine;hydrochloride
SMILES C1=CC(=CC=C1NN)[N+](=O)[O-].Cl

Specifications

Quantity 5 g
Formula Weight 189.60
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylhydrazine Hydrochloride
EINECSNumber (3)-0477
TSCA Yes