missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylboronic acid, 97%, Thermo Scientific™

$154.33 - $463.86

Chemical Identifiers

CAS 24067-17-2
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161360
InChI Key NSFJAFZHYOAMHL-UHFFFAOYSA-N
Synonym 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
PubChem CID 2773552
IUPAC Name (4-nitrophenyl)boronic acid
SMILES B(C1=CC=C(C=C1)[N+](=O)[O-])(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC433890010
View Documents Promotion Details
N/A 1g Glass bottle
Each for $154.33
Only null left
 
AC433890050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $463.86
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

24067-17-2
166.93
NSFJAFZHYOAMHL-UHFFFAOYSA-N
2773552
B(C1=CC=C(C=C1)[N+](=O)[O-])(O)O
C6H6BNO4
MFCD00161360
4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
(4-nitrophenyl)boronic acid

Specifications

24067-17-2
100.0
96% min. (HPLC)
MFCD00161360
NSFJAFZHYOAMHL-UHFFFAOYSA-N
(4-nitrophenyl)boronic acid
2773552
97%
96.0
Authentic
C6H6BNO4
4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
B(C1=CC=C(C=C1)[N+](=O)[O-])(O)O
166.93
166.93
4-Nitrophenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only