missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7133689
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
N022725G 25 g
N0227500G 500 g
2 options

Catalog No. N022725G

Supplier: TCI America N022725G

Only null left

Chemical Identifiers

CAS 104-03-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007383
InChI Key YBADLXQNJCMBKR-UHFFFAOYSA-N
Synonym 4-nitrophenylacetic acid, 2-4-nitrophenyl acetic acid, p-nitrophenylacetic acid, 4-nitrophenyl acetic acid, 4-nitrobenzeneacetic acid, benzeneacetic acid, 4-nitro, 2-p-nitrophenyl acetic acid, 4-nitrophenylacetate, 4-nitrophenylaceticacid, acetic acid, p-nitrophenyl
PubChem CID 4661
ChEBI CHEBI:40443
SMILES C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylacetic Acid
EINECSNumber (3)-1749
RTECSNumber AJ1130010
TSCA Yes