missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetic acid, 98%

CAS: 104-03-0 | C8H7NO4 | 181.147 g/mol

Supplier:  Thermo Scientific Chemicals A1590136

 View more versions of this product

Catalog No. AAA1590136


Only null left
Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis and also used for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis and also used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
Specifications

Specifications

4-Nitrophenylacetic acid
152°C to 156°C
MFCD00007383
1911801
4-nitrophenylacetic acid, 2-4-nitrophenyl acetic acid, p-nitrophenylacetic acid, 4-nitrophenyl acetic acid, 4-nitrobenzeneacetic acid, benzeneacetic acid, 4-nitro, 2-p-nitrophenyl acetic acid, 4-nitrophenylacetate, 4-nitrophenylaceticacid, acetic acid, p-nitrophenyl
YBADLXQNJCMBKR-UHFFFAOYSA-N
181.147
CHEBI:40443
98%
104-03-0
C8H7NO4
500 g
14,6622
Slightly soluble in water.
C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]
4661
181.15

Chemical Identifiers

104-03-0
181.147
YBADLXQNJCMBKR-UHFFFAOYSA-N
4661
C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]
C8H7NO4
MFCD00007383
4-nitrophenylacetic acid, 2-4-nitrophenyl acetic acid, p-nitrophenylacetic acid, 4-nitrophenyl acetic acid, 4-nitrobenzeneacetic acid, benzeneacetic acid, 4-nitro, 2-p-nitrophenyl acetic acid, 4-nitrophenylacetate, 4-nitrophenylaceticacid, acetic acid, p-nitrophenyl
CHEBI:40443
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 203-168-5

RTECSNumber : AJ1130010

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only