missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylacetic acid, 98%

Catalog No. AAA1590114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590114 25 g
AAA1590122 100 g
AAA1590136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1590114 Supplier Thermo Scientific Chemicals Supplier No. A1590114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104-03-0 | C8H7NO4 | 181.147 g/mol

It is an important raw material and intermediate used in organic synthesis and also used for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis and also used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 104-03-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007383
InChI Key YBADLXQNJCMBKR-UHFFFAOYSA-N
Synonym 4-nitrophenylacetic acid, 2-4-nitrophenyl acetic acid, p-nitrophenylacetic acid, 4-nitrophenyl acetic acid, 4-nitrobenzeneacetic acid, benzeneacetic acid, 4-nitro, 2-p-nitrophenyl acetic acid, 4-nitrophenylacetate, 4-nitrophenylaceticacid, acetic acid, p-nitrophenyl
PubChem CID 4661
ChEBI CHEBI:40443
SMILES C1=CC(=CC=C1CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point 152°C to 156°C
Quantity 25 g
Beilstein 1911801
Merck Index 14,6622
Solubility Information Slightly soluble in water.
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-168-5
RTECSNumber AJ1130010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only