missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrophenyl)thiourea 98.0+%, TCI America™

Catalog No. N09951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09951G Supplier TCI America Supplier No. N09951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3696-22-8
Molecular Formula C7H7N3O2S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00007304
InChI Key BLYAANPIHFKKMQ-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl-2-thiourea, n-4-nitrophenyl thiourea, 4-nitrophenyl thiourea, 1-4-nitrophenyl thiourea, p-nitrophenyl thiourea, thiourea, 4-nitrophenyl, n-p-nitrophenyl thiourea, acmc-1cny6, 4-nitro-phenyl-thiourea, n-4-nitrophenyl thiourea #
PubChem CID 2760234
IUPAC Name (4-nitrophenyl)thiourea
SMILES NC(=S)NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 206°C
Quantity 1 g
UN Number 2811
Formula Weight 197.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Nitrophenyl)thiourea