Learn More
4-Nitrophenyl phosphate, 97%
CAS: 330-13-2 | C6H6NO6P | 219.09 g/mol
Supplier: Thermo Scientific Chemicals 455010100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Nitrophenyl phosphate | |
| Authentic | |
| C6H6NO6P | |
| 10 g | |
| XZKIHKMTEMTJQX-UHFFFAOYSA-N | |
| 219.09 | |
| CHEBI:17440 | |
| 97% |
| 330-13-2 | |
| 96% min. (HPLC) | |
| O2NC6H4OP(O)(OH)2 | |
| 4-nitrophenyl phosphate, 4-nitrophenyl dihydrogen phosphate, nitrophenylphosphate, phosphoric acid, mono 4-nitrophenyl ester, p-nitrophenyl phosphate, p-nitrophenyl dihydrogen phosphate, p-nitrophenol phosphate, p-nitrophenylphosphate, pnpp | |
| C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O | |
| 378 | |
| 219.09 |
Chemical Identifiers
| 330-13-2 | |
| 219.09 | |
| 4-nitrophenyl phosphate, 4-nitrophenyl dihydrogen phosphate, nitrophenylphosphate, phosphoric acid, mono 4-nitrophenyl ester, p-nitrophenyl phosphate, p-nitrophenyl dihydrogen phosphate, p-nitrophenol phosphate, p-nitrophenylphosphate, pnpp | |
| CHEBI:17440 |
| C6H6NO6P | |
| XZKIHKMTEMTJQX-UHFFFAOYSA-N | |
| 378 | |
| C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 206-353-9
RUO – Research Use Only