missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate, 97%

Catalog No. p-7077528
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455010100 10 g
AC455010500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455010100 Supplier Thermo Scientific Chemicals Supplier No. 455010100
Only null left

CAS: 330-13-2 | C6H6NO6P | 219.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330-13-2
Molecular Formula C6H6NO6P
Molecular Weight (g/mol) 219.09
InChI Key XZKIHKMTEMTJQX-UHFFFAOYSA-N
Synonym 4-nitrophenyl phosphate, 4-nitrophenyl dihydrogen phosphate, nitrophenylphosphate, phosphoric acid, mono 4-nitrophenyl ester, p-nitrophenyl phosphate, p-nitrophenyl dihydrogen phosphate, p-nitrophenol phosphate, p-nitrophenylphosphate, pnpp
PubChem CID 378
ChEBI CHEBI:17440
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)(O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Linear Formula O2NC6H4OP(O)(OH)2
Quantity 10 g
Formula Weight 219.09
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl phosphate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 206-353-9

RUO – Research Use Only