missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl palmitate, 98+%

Catalog No. p-7045483
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1089603 1 g
AAL1089606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1089603 Supplier Thermo Scientific Chemicals Supplier No. L1089603
Only null left

CAS: 1492-30-4 | C22H35NO4 | 377.525 g/mol

The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.

Solubility
Insoluble in water. Soluble in CHCl3.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C

Chemical Identifiers

CAS 1492-30-4
Molecular Formula C22H35NO4
Molecular Weight (g/mol) 377.525
MDL Number MFCD00047732
InChI Key LVZSQWIWCANHPF-UHFFFAOYSA-N
Synonym 4-nitrophenyl palmitate, p-nitrophenyl palmitate, hexadecanoic acid 4-nitrophenyl ester, 4-nitrophenyl hexadecanoate, hexadecanoic acid, 4-nitrophenyl ester, 4-nitrophenylpalmitate, paranitrophenyl palmitate, para-nitrophenyl palmitate, p-nitrophenyl hexadecanoate, 4-nitrophenyl palmitate #
PubChem CID 73891
ChEBI CHEBI:85645
IUPAC Name (4-nitrophenyl) hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 65°C to 66°C
Quantity 1 g
Beilstein 1891754
Solubility Information Insoluble in water. Soluble in CHCl3.
Formula Weight 377.53
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl palmitate
EINECSNumber 216-084-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only