missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl N-Benzylcarbamate 98.0+%, TCI America™

Catalog No. p-7134060
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0978200MG 200 mg
N09781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0978200MG Supplier TCI America Supplier No. N0978200MG

Chemical Identifiers

CAS 124068-97-9
Molecular Formula C14H12N2O4
Molecular Weight (g/mol) 272.26
MDL Number MFCD06796634
InChI Key OCAOEODBVXZHNZ-UHFFFAOYSA-N
Synonym N-Benzylcarbamic Acid 4-Nitrophenyl Ester
PubChem CID 183685
IUPAC Name (4-nitrophenyl) N-benzylcarbamate
SMILES C1=CC=C(C=C1)CNC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 132°C
Quantity 200 mg
Formula Weight 272.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl N-Benzylcarbamate