CAS RN 124068-97-9
IUPAC Name:
4-nitrophenyl N-benzylcarbamate
Synonyms:
4-nitrophenyl N-benzylcarbamate
Molecular Weight (g/mol):
272.26
Molecular Formula:
C14H12N2O4
InChi Key:
OCAOEODBVXZHNZ-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(OC(=O)NCC2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
4-Nitrophenyl N-Benzylcarbamate 98.0+%, TCI America™
CAS: 124068-97-9 Molecular Formula: C14H12N2O4 Molecular Weight (g/mol): 272.26 MDL Number: MFCD06796634 InChI Key: OCAOEODBVXZHNZ-UHFFFAOYSA-N Synonym: N-Benzylcarbamic Acid 4-Nitrophenyl Ester PubChem CID: 183685 IUPAC Name: (4-nitrophenyl) N-benzylcarbamate SMILES: C1=CC=C(C=C1)CNC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]