missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophényl N-Benzylcarbamate 98,0+%, TCI America™

Catalog No. N0978200MG
Change view
Click to view available options
Quantité:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
N0978200MG 200 mg
N09781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0978200MG Supplier TCI America Supplier No. N0978200MG

Chemical Identifiers

CAS 124068-97-9
Formule moléculaire C14H12N2O4
Poids moléculaire (g/mol) 272.26
Numéro MDL MFCD06796634
Clé InChI OCAOEODBVXZHNZ-UHFFFAOYSA-N
Synonyme N-Benzylcarbamic Acid 4-Nitrophenyl Ester
PubChem CID 183685
Nom de l’IUPAC (4-nitrophényl) N-benzylcarbamate
SOURIRES C1=CC=C(C=C1)CNC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Nom ou substance chimique 4-Nitrophenyl N-Benzylcarbamate
Point de fusion 132°C
Quantité 200 mg
Poids de formule 272.26
Pourcentage de pureté ≥98.0% (N)
Forme physique Poudre cristalline