missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Laurate 98.0+%, TCI America™

Catalog No. L01001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01001G 1 g
L01005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01001G Supplier TCI America Supplier No. L01001G
Only null left

Chemical Identifiers

CAS 1956-11-2
Molecular Formula C18H27NO4
Molecular Weight (g/mol) 321.42
MDL Number MFCD00047730
InChI Key YNGNVZFHHJEZKD-UHFFFAOYSA-N
Synonym Lauric Acid 4-Nitrophenyl Ester
PubChem CID 74778
IUPAC Name (4-nitrophenyl) dodecanoate
SMILES CCCCCCCCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 45°C
Color White
Quantity 1 g
Formula Weight 321.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Laurate