missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl Isothiocyanate 99.0+%, TCI America™
Supplier: TCI America N082625G
Specifications
| 4-Nitrophenyl Isothiocyanate | |
| 112°C | |
| 138°C | |
| MFCD00007307 | |
| 2811 | |
| NXHSSIGRWJENBH-UHFFFAOYSA-N | |
| 1-isothiocyanato-4-nitrobenzene | |
| 75044 | |
| ≥99.0% (GC) |
| 2131-61-5 | |
| Yellow | |
| C7H4N2O2S | |
| 25 g | |
| 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate | |
| C1=CC(=CC=C1N=C=S)[N+](=O)[O-] | |
| 180.181 | |
| 180.18 | |
| Crystalline Powder |
Chemical Identifiers
| 2131-61-5 | |
| 180.181 | |
| NXHSSIGRWJENBH-UHFFFAOYSA-N | |
| 75044 | |
| C1=CC(=CC=C1N=C=S)[N+](=O)[O-] |
| C7H4N2O2S | |
| MFCD00007307 | |
| 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate | |
| 1-isothiocyanato-4-nitrobenzene |
Safety and Handling
TSCA : Yes