missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl isothiocyanate, 97%

Catalog No. AAL1104209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1104209 10 g
AAL1104218 50 g
AAL1104230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1104209 Supplier Thermo Scientific Chemicals Supplier No. L1104209

CAS: 2131-61-5 | C7H4N2O2S | 180.181 g/mol

4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production

Solubility
It hydrolyzes with water. Soluble in toluene.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 2131-61-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD00007307
InChI Key NXHSSIGRWJENBH-UHFFFAOYSA-N
Synonym 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
PubChem CID 75044
IUPAC Name 1-isothiocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=S)[N+](=O)[O-]

Specifications

Melting Point 105°C to 110°C
Boiling Point 137°C to 138°C (11 mmHg)
Odor Irritating
Quantity 10 g
UN Number UN2923
Beilstein 640027
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water. Soluble in toluene.
Formula Weight 180.19
Percent Purity 97%
Chemical Name or Material 4-Nitrophenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-359-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only