missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Hexanoate 98.0+%, TCI America™

Catalog No. H04841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04841G 1 g
H04845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04841G Supplier TCI America Supplier No. H04841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 956-75-2
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00056111
InChI Key OLRXUEYZKCCEKK-UHFFFAOYSA-N
Synonym Hexanoic Acid 4-Nitrophenyl Ester
PubChem CID 567162
IUPAC Name (4-nitrophenyl) hexanoate
SMILES CCCCCC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Boiling Point 145°C
Quantity 1 g
Formula Weight 237.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Nitrophenyl Hexanoate
TSCA No
Recommended Storage Refrigerator