missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl chloroformate, 97%

CAS: 7693-46-1 | C7H4ClNO4 | 201.57 g/mol

Supplier:  Thermo Scientific Chemicals 170800250

 View more versions of this product

Catalog No. AC170800250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

4-Nitrophenyl chloroformate
96.0
73.0°C to 81.0°C
159.0°C to 162.0°C (19.0 mmHg)
Authentic
Glass bottle
ClCO2C6H4NO2
25 g
02,297
Solubility in water: decomposes. Other solubilities: soluble in toluene and benzene
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl
201.57
201.57
Crystalline Powder, Crystals or Chunks
7693-46-1
100.0
Beige-Yellow to White
>110°C
96% min. (ex Cl) (Argentometry)
C7H4ClNO4
MFCD00007321
06, I, 120
4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
NXLNNXIXOYSCMB-UHFFFAOYSA-N
(4-nitrophenyl) carbonochloridate
82129
97%

Chemical Identifiers

7693-46-1
201.57
NXLNNXIXOYSCMB-UHFFFAOYSA-N
82129
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl
C7H4ClNO4
MFCD00007321
4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
(4-nitrophenyl) carbonochloridate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-706-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only