missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl chloroformate, 97%

Catalog No. AC170801000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170801000 100 g Glass bottle
AC170800050 5 g Glass bottle
AC170800250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC170801000 Supplier Thermo Scientific Chemicals Supplier No. 170801000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7693-46-1 | C7H4ClNO4 | 201.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 7693-46-1
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.57
MDL Number MFCD00007321
InChI Key NXLNNXIXOYSCMB-UHFFFAOYSA-N
Synonym 4-nitrophenyl chloroformate, carbonochloridic acid, 4-nitrophenyl ester, chloroformic acid 4-nitrophenyl ester, p-nitrophenyl chloroformate, chloroformic acid p-nitrophenyl ester, 4-nitrophenyl carbonochloridate, formic acid, chloro-, p-nitrophenyl ester, 4-nitrophenylchlorocarbonat, 4-nitrophenyl choroformate
PubChem CID 82129
IUPAC Name (4-nitrophenyl) carbonochloridate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73.0°C to 81.0°C
Color Beige-Yellow to White
Boiling Point 159.0°C to 162.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula ClCO2C6H4NO2
Quantity 100 g
Beilstein 06, I, 120
Fieser 02,297
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in toluene and benzene
Formula Weight 201.57
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Nitrophenyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 231-706-9

RUO – Research Use Only