Learn More
4-Nitrophenyl beta-D-galactopyranoside, 98+%
CAS: 3150-24-1 | C12H15NO8 | 301.25 g/mol
Supplier: Thermo Scientific Chemicals H5690506
Description
It is useful in detecting the glycosylated enzyme.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is useful in detecting the glycosylated enzyme.
Solubility
Soluble in  DMSO, Methanol, Water.
Notes
Store at -20°C. Incompatible with oxidizing agents and heat.
Specifications
| 4-Nitrophenyl beta-D-galactopyranoside | |
| C12H15NO8 | |
| 5 g | |
| 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside | |
| IFBHRQDFSNCLOZ-YBXAARCKSA-N | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol | |
| 65115 | |
| 301.25 |
| 3150-24-1 | |
| MFCD00063256 | |
| 92213 | |
| Soluble in DMSO,Methanol,Water. | |
| OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O | |
| 301.25 | |
| CHEBI:355715 | |
| ≥98% |
Chemical Identifiers
| 3150-24-1 | |
| 301.25 | |
| IFBHRQDFSNCLOZ-YBXAARCKSA-N | |
| 65115 | |
| (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| C12H15NO8 | |
| MFCD00063256 | |
| 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside | |
| CHEBI:355715 | |
| OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O |
Safety and Handling
EINECSNumber : 221-584-5
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only