missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl beta-D-galactopyranoside, 98+%

Catalog No. p-7038033
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5690506 5 g
AAH5690509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5690506 Supplier Thermo Scientific Chemicals Supplier No. H5690506

CAS: 3150-24-1 | C12H15NO8 | 301.25 g/mol

It is useful in detecting the glycosylated enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is useful in detecting the glycosylated enzyme.

Solubility
Soluble in  DMSO, Methanol, Water.

Notes
Store at -20°C. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 3150-24-1
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00063256
InChI Key IFBHRQDFSNCLOZ-YBXAARCKSA-N
Synonym 4-nitrophenyl-beta-d-galactopyranoside, 4-nitrophenylgalactoside, p-nitrophenyl-beta-d-galactoside, p-nitrophenyl beta-d-galactopyranoside, 4-nitrophenyl beta-d-galactopyranoside, p-nitrophenol galactoside, 4-nitrophenyl-beta-d-galactoside, 4-nitrophenyl galactoside, 1-o-p-nitrophenyl-beta-d-galactopyranose, 4-nitrophenylbeta-d-galactopyranoside
PubChem CID 65115
ChEBI CHEBI:355715
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 5 g
Beilstein 92213
Solubility Information Soluble in DMSO,Methanol,Water.
Formula Weight 301.25
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl beta-D-galactopyranoside
EINECSNumber 221-584-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only