missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl beta-D-Fucopyranoside 98.0+%, TCI America™

Catalog No. N0774100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0774100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0774100MG Supplier TCI America Supplier No. N0774100MG
Only null left

Chemical Identifiers

CAS 1226-39-7
Molecular Formula C12H15NO7
Molecular Weight (g/mol) 285.252
InChI Key YILIDCGSXCGACV-BVWHHUJWSA-N
Synonym 4-nitrophenyl beta-d-fucopyranoside, 4-nitrophenyl b-d-fucopyranoside, 4-nitrophenyl-beta-d-fucopyranoside, p-nitrophenyl beta-d-fucopyranoside, 2r,3r,4s,5r,6s-2-methyl-6-4-nitrophenoxy oxane-3,4,5-triol, b-d-galactopyranoside,4-nitrophenyl 6-deoxy, 4-nitrophenyl 6-deoxy-beta-d-galactopyranoside, 4-nitrophenyl, a-d-fucopyranoside, 2-nitrophenyl i 2-d-fucopyranoside, 4-nitrophenyl i 2-d-fucopyranoside
PubChem CID 2733769
IUPAC Name (2R,3R,4S,5R,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES CC1C(C(C(C(O1)OC2=CC=C(C=C2)[N+](=O)[O-])O)O)O

Specifications

Melting Point 184°C
Color White
Quantity 100 mg
Formula Weight 285.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl beta-D-Fucopyranoside