accessibility menu, dialog, popup

UserName

CAS RN 1226-39-7

CH3(R)O(S)ONOO(R)OH(S)HO(R)HO

CAS RN 1226-39-7

IUPAC Name: (2R,3R,4S,5R,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms: (2R,3R,4S,5R,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 285.25
Molecular Formula: C12H15NO7
InChi Key: YILIDCGSXCGACV-BVWHHUJWSA-N
SMILES: C[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results