missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-alpha-L-Fucopyranoside

Catalog No. aaj3413103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ3413103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ3413103 Supplier Thermo Fisher Scientific Supplier No. J3413103
Only null left

CAS: 10231-84-2285.252 g/mol

 

Chemical Identifiers

CAS 10231-84-2
Molecular Weight (g/mol) 285.252
Synonym 4-nitrophenyl a-l-fucopyranoside

Specifications

Physical Form Powder
Quantity 1 g
Packaging Plastic Bottle
Chemical Name or Material 4-Nitrophenyl-alpha-L-Fucopyranoside
Recommended Storage Keep cold

RUO – Research Use Only