accessibility menu, dialog, popup

UserName

CH3(S)O(S)ONOO(S)OH(R)HO(S)HO

CAS RN 10231-84-2

IUPAC Name: (2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms: (2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 285.25
Molecular Formula: C12H15NO7
InChi Key: YILIDCGSXCGACV-SQKFTNEHSA-N
SMILES: C[C@@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@@H](O)[C@H](O)[C@@H]1O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results