missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl alpha-D-mannopyranoside, 98%

CAS: 10357-27-4 | C12H15NO8 | 301.251 g/mol

Supplier:  Thermo Scientific Chemicals J6602603

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6602603


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10357-27-4
301.251
IFBHRQDFSNCLOZ-GCHJQGSQSA-N
449023
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
C12H15NO8
MFCD00066002
4-nitrophenyl a-d-mannopyranoside, 4-nitrophenyl alpha-d-mannopyranoside, 4-nitrophenyl-alpha-d-mannopyranoside, a-d-mannopyranoside, 4-nitrophenyl, p-nitrophenyl-alpha-d-mannopyranoside, 4'-nitrophenyl-alpha-d-mannopyranoside, 2r,3s,4s,5s,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, pnpalphaman, pnp-alpha-d-man, 4-nitrophenyla-d-mannopyranoside
(2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol

Specifications

4-Nitrophenyl alpha-D-mannopyranoside
178°C to 183°C
C12H15NO8
MFCD00066002
4-nitrophenyl a-d-mannopyranoside, 4-nitrophenyl alpha-d-mannopyranoside, 4-nitrophenyl-alpha-d-mannopyranoside, a-d-mannopyranoside, 4-nitrophenyl, p-nitrophenyl-alpha-d-mannopyranoside, 4'-nitrophenyl-alpha-d-mannopyranoside, 2r,3s,4s,5s,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, pnpalphaman, pnp-alpha-d-man, 4-nitrophenyla-d-mannopyranoside
IFBHRQDFSNCLOZ-GCHJQGSQSA-N
(2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
449023
98%
10357-27-4
White
1 g
92210
Soluble in water at 10mg/ml
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
301.251
301.25
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-776-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions