missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl alpha-D-mannopyranoside, 98%

Catalog No. p-7043184
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66026ME 500 mg
AAJ6602603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66026ME Supplier Thermo Scientific Chemicals Supplier No. J66026ME

CAS: 10357-27-4 | C12H15NO8 | 301.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10357-27-4
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00066002
InChI Key IFBHRQDFSNCLOZ-GCHJQGSQSA-N
Synonym 4-nitrophenyl a-d-mannopyranoside, 4-nitrophenyl alpha-d-mannopyranoside, 4-nitrophenyl-alpha-d-mannopyranoside, a-d-mannopyranoside, 4-nitrophenyl, p-nitrophenyl-alpha-d-mannopyranoside, 4'-nitrophenyl-alpha-d-mannopyranoside, 2r,3s,4s,5s,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, pnpalphaman, pnp-alpha-d-man, 4-nitrophenyla-d-mannopyranoside
PubChem CID 449023
IUPAC Name (2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 178°C to 183°C
Color White
Quantity 500 mg
Beilstein 92210
Solubility Information Soluble in water at 10mg/ml
Formula Weight 301.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Nitrophenyl alpha-D-mannopyranoside
EINECSNumber 233-776-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only