missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl alpha-D-Glucopyranoside 98.0+%, TCI America™
$94.93 - $320.87
Chemical Identifiers
| CAS | 3767-28-0 |
|---|---|
| Molecular Formula | C12H15NO8 |
| Molecular Weight (g/mol) | 301.251 |
| MDL Number | MFCD00064088 |
| InChI Key | IFBHRQDFSNCLOZ-ZIQFBCGOSA-N |
| Synonym | 4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside |
| PubChem CID | 92969 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| SMILES | C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O |
Chemical Identifiers
| C12H15NO8 | |
| MFCD00064088 | |
| 4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol |
Specifications
| 3767-28-0 | |
| Yellow | |
| MFCD00064088 | |
| IFBHRQDFSNCLOZ-ZIQFBCGOSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol | |
| 92969 | |
| ≥98.0% (HPLC) | |
| 4-Nitrophenyl alpha-D-Glucopyranoside [Substrate for alpha-D-Glucosidase] |
| 212°C | |
| C12H15NO8 | |
| 4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside | |
| C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O | |
| 301.251 | |
| 301.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator