missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl alpha-D-Glucopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 212°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 301.25 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrophenyl alpha-D-Glucopyranoside [Substrate for alpha-D-Glucosidase] |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |