missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside 98.0+%, TCI America™

$125.31 - $377.82

Chemical Identifiers

CAS 3459-18-5
Molecular Formula C14H18N2O8
Molecular Weight (g/mol) 342.30
MDL Number MFCD00063696
InChI Key OMRLTNCLYHKQCK-JQXIRTQVNA-N
Synonym 4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
PubChem CID 102416
IUPAC Name N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
N0866200MG
View Documents Promotion Details
N/A 200 mg
Each for $125.31
Only null left
 
N08661G
View Documents Promotion Details
N/A 1 g
Each for $377.82
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3459-18-5
342.30
OMRLTNCLYHKQCK-JQXIRTQVNA-N
102416
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O
C14H18N2O8
MFCD00063696
4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide

Specifications

3459-18-5
White-Yellow
MFCD00063696
OMRLTNCLYHKQCK-JQXIRTQVNA-N
N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
102416
≥98.0% (HPLC,N)
4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
205°C
C14H18N2O8
4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O
342.30
342.30
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents