missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenoxyacetic Acid 98.0+%, TCI America™

Catalog No. N022625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N022625G 25 g
N0226500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N022625G Supplier TCI America Supplier No. N022625G
Only null left

Chemical Identifiers

CAS 1798-11-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00017030
InChI Key AVDLFIONKHGQAP-UHFFFAOYSA-N
Synonym 4-nitrophenoxyacetic acid, 2-4-nitrophenoxy acetic acid, 4-nitrophenoxy acetic acid, p-nitrophenoxyacetic acid, acetic acid, 4-nitrophenoxy, 4-nitro-phenoxy-acetic acid, acetic acid, p-nitrophenoxy, acetic acid, 2-4-nitrophenoxy, acmc-209efx
PubChem CID 15720
SMILES C1=CC(=CC=C1[N+](=O)[O-])OCC(=O)O

Specifications

Melting Point 187°C
Quantity 25 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenoxyacetic Acid
EINECSNumber (3)-0781
RTECSNumber AJ1100000
TSCA Yes