missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitroindole 98.0+%, TCI America™
$65.24 - $190.13
Chemical Identifiers
| CAS | 4769-97-5 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.148 |
| MDL Number | MFCD00010056 |
| InChI Key | LAVZKLJDKGRZJG-UHFFFAOYSA-N |
| Synonym | 4-nitroindole, indole, 4-nitro, 1h-indole, 4-nitro, pubchem7440, 4-nitro-1 h-indole, #, maybridge1_006408, acmc-209ka5, ksc236o2l |
| PubChem CID | 145766 |
| IUPAC Name | 4-nitro-1H-indole |
| SMILES | C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-] |
Chemical Identifiers
| 4769-97-5 | |
| 162.148 | |
| LAVZKLJDKGRZJG-UHFFFAOYSA-N | |
| 145766 | |
| C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-] |
| C8H6N2O2 | |
| MFCD00010056 | |
| 4-nitroindole, indole, 4-nitro, 1h-indole, 4-nitro, pubchem7440, 4-nitro-1 h-indole, #, maybridge1_006408, acmc-209ka5, ksc236o2l | |
| 4-nitro-1H-indole |
Specifications
| 4769-97-5 | |
| C8H6N2O2 | |
| 1 g | |
| LAVZKLJDKGRZJG-UHFFFAOYSA-N | |
| 4-nitro-1H-indole | |
| 145766 | |
| ≥98.0% (GC) | |
| 4-Nitroindole |
| 206°C | |
| MFCD00010056 | |
| 4-nitroindole, indole, 4-nitro, 1h-indole, 4-nitro, pubchem7440, 4-nitro-1 h-indole, #, maybridge1_006408, acmc-209ka5, ksc236o2l | |
| C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-] | |
| 162.148 | |
| 162.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No