missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrodiphenylamine, 98+%

Catalog No. AAL0072803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0072803 1 g
AAL0072806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0072803 Supplier Thermo Scientific Chemicals Supplier No. L0072803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 836-30-6 | C12H10N2O2 | 214.22 g/mol

4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrodiphenylamine, is used as a Hammett indicator. It is also used as an Intermediate in the production of rubber antioxidants. It is manufactured solely for use as a chemical intermediate for industrial uses and thus is essentially fully consumed in chemical reductive alkylation reaction downstream of manufacture.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 836-30-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00007301
InChI Key XXYMSQQCBUKFHE-UHFFFAOYSA-N
Synonym 4-nitrodiphenylamine, p-nitrodiphenylamine, benzenamine, 4-nitro-n-phenyl, diphenylamine, 4-nitro, 4-nitrodifenylamin, 4-ndpa, p-nitrophenylphenylamine, 4-nitro-n-phenyl-aniline, 4-nitrophenyl phenylamine, 4-nitrodifenylamin czech
PubChem CID 13271
IUPAC Name 4-nitro-N-phenylaniline
SMILES [O-][N+](=O)C1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 132°C to 135°C
Boiling Point 211°C (30 mmHg)
Flash Point 190°C (374°F)
Quantity 1 g
UN Number UN3077
Beilstein 2051910
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity ≥98%
Chemical Name or Material 4-Nitrodiphenylamine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 212-646-2
RTECSNumber JJ9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only