missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinnamyl Alcohol 98.0+%, TCI America™

Catalog No. p-7133939
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06901G 1 g
N06905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06901G Supplier TCI America Supplier No. N06901G
Only null left

Chemical Identifiers

CAS 1504-63-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00017045
InChI Key LGXXEDSIJZHDBN-OWOJBTEDSA-N
Synonym 4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
PubChem CID 5462894
IUPAC Name (E)-3-(4-nitrophenyl)prop-2-en-1-ol
SMILES C1=CC(=CC=C1C=CCO)[N+](=O)[O-]

Specifications

Melting Point 128°C
Quantity 1 g
Formula Weight 179.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrocinnamyl Alcohol