missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrochalcone 95.0+%, TCI America™
Supplier: TCI America N083925G
Specifications
| 4-Nitrochalcone | |
| 165°C | |
| C15H11NO3 | |
| 25 g | |
| WDZGGAFMGIOIQS-DHZHZOJOSA-N | |
| (E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one | |
| 5377323 | |
| ≥95.0% (GC) |
| 1222-98-6 | |
| Yellow | |
| MFCD00007382 | |
| 4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669 | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-] | |
| 253.257 | |
| 253.26 | |
| Crystalline Powder |
Chemical Identifiers
| 1222-98-6 | |
| 253.257 | |
| WDZGGAFMGIOIQS-DHZHZOJOSA-N | |
| 5377323 | |
| C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-] |
| C15H11NO3 | |
| MFCD00007382 | |
| 4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669 | |
| (E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one |
Safety and Handling
RTECSNumber : FL7075500
TSCA : Yes