missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrochalcone 95.0+%, TCI America™

Catalog No. p-7133992
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08395G 5 g
N083925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08395G Supplier TCI America Supplier No. N08395G
Only null left

Chemical Identifiers

CAS 1222-98-6
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00007382
InChI Key WDZGGAFMGIOIQS-DHZHZOJOSA-N
Synonym 4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
PubChem CID 5377323
IUPAC Name (E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 165°C
Color Yellow
Quantity 5 g
Formula Weight 253.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrochalcone
RTECSNumber FL7075500
TSCA Yes