missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocatechol 98.0+%, TCI America™

Catalog No. p-7133719
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02735G 5 g
N027325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02735G Supplier TCI America Supplier No. N02735G
Only null left

Chemical Identifiers

CAS 3316-09-4
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00007242
InChI Key XJNPNXSISMKQEX-UHFFFAOYSA-N
Synonym 4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc
PubChem CID 3505109
ChEBI CHEBI:16318
IUPAC Name 4-nitrobenzene-1,2-diol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)O

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 155.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrocatechol