missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Bromide 98.0+%, TCI America™

Supplier:  TCI America N0181500G

 View more versions of this product

Catalog No. N0181500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Nitrobenzyl Bromide
99°C
MFCD00007373
3261
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
1-(bromomethyl)-4-nitrobenzene
66011
216.03
Crystalline Powder
100-11-8
C7H6BrNO2
500 g
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
[O-][N+](=O)C1=CC=C(CBr)C=C1
216.03
CHEBI:62822
≥98.0% (GC,T)

Chemical Identifiers

100-11-8
216.03
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
66011
1-(bromomethyl)-4-nitrobenzene
C7H6BrNO2
MFCD00007373
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
CHEBI:62822
[O-][N+](=O)C1=CC=C(CBr)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : XS7967000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions