missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Bromide 98.0+%, TCI America™

Catalog No. N018125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N018125G 25 g
N0181100G 100 g
N0181500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N018125G Supplier TCI America Supplier No. N018125G

Chemical Identifiers

CAS 100-11-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007373
InChI Key VOLRSQPSJGXRNJ-UHFFFAOYSA-N
Synonym 4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
PubChem CID 66011
ChEBI CHEBI:62822
IUPAC Name 1-(bromomethyl)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 99°C
Quantity 25 g
UN Number 3261
Formula Weight 216.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Bromide
RTECSNumber XS7967000
TSCA Yes