missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl bromide, 99%

CAS: 100-11-8 | C7H6BrNO2 | 216.03 g/mol

$84.05 - $298.55

Chemical Identifiers

CAS 100-11-8
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007373
InChI Key VOLRSQPSJGXRNJ-UHFFFAOYSA-N
Synonym 4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
PubChem CID 66011
ChEBI CHEBI:62822
IUPAC Name 1-(bromomethyl)-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(CBr)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC128530250
SDS View Documents
Thermo Scientific Chemicals
128530250
25 g
Each for $84.05
Only null left
 
AC128531000
SDS View Documents
Thermo Scientific Chemicals
128531000
100 g
Each for $298.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-11-8
216.03
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
66011
1-(bromomethyl)-4-nitrobenzene
C7H6BrNO2
MFCD00007373
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
CHEBI:62822
[O-][N+](=O)C1=CC=C(CBr)C=C1

Specifications

100-11-8
Yellow to Beige
Authentic
Glass bottle
O2NC6H4CH2Br
25 g
01,736; 08,36
Solubility in water: insoluble. Other solubilities: soluble in dmf,isopropyl acetate and acetone
[O-][N+](=O)C1=CC=C(CBr)C=C1
216.03
CHEBI:62822
99%
4-Nitrobenzyl bromide
98.0°C to 100.0°C
>100°C
98.5% min. (GC)
C7H6BrNO2
MFCD00007373
05,334
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
1-(bromomethyl)-4-nitrobenzene
66011
216.03
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 202-820-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only