missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl alcohol, 99%

Catalog No. p-7022819
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1574214 25 g
AAA1574222 100 g
AAA1574236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1574214 Supplier Thermo Scientific Chemicals Supplier No. A1574214

CAS: 619-73-8 | C7H7NO3 | 153.14 g/mol

4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrobenzyl alcohol is used as the sole source of carbon and nitrogen to study the pathway for the catabolism of 4-nitrotoluene by Pseudomonas.

Solubility
Soluble in water (2 mg/ml at 20°C).

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 619-73-8
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007376
InChI Key JKTYGPATCNUWKN-UHFFFAOYSA-N
Synonym 4-nitrobenzyl alcohol, 4-nitrophenyl methanol, p-nitrobenzyl alcohol, benzenemethanol, 4-nitro, 4-nitrobenzenemethanol, p-hydroxymethyl nitrobenzene, benzyl alcohol, p-nitro, paranitrobenzyl alcohol, 4-nitrobenzylalcohol, unii-86btj68y9m
PubChem CID 69275
ChEBI CHEBI:41214
IUPAC Name (4-nitrophenyl)methanol
SMILES OCC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 92°C to 95°C
Boiling Point 185°C (12 mmHg)
Flash Point 180°C (356°F)
Quantity 25 g
Beilstein 1424026
Solubility Information Soluble in water (2mg/ml at 20°C).
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-611-6
RTECSNumber DP0657100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only